Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pbx4
Entrez Gene ID
361131
Gene Name
pre-B-cell leukemia homeobox 4
Gene Aliases
Edg4
Chromosome Location
Chr.16: 21303117 - 21338771 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108399.1 NP_001101869.1 3 - 4 497 67
XM_006252909.2 XP_006252971.1 4 - 5 488 67
XM_008771129.1 XP_008769351.1 3 - 4 592 67
XM_008771130.1 XP_008769352.1 3 - 4 350 67

View Full Product Details