Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankle1
Entrez Gene ID
361122
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
Ankrd41, RGD1308184
Chromosome Location
Chr.16: 19800390 - 19826057 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252959.2 XP_006253021.1 3 - 4 393 93
XM_006252960.2 XP_006253022.1 3 - 4 392 93
XM_006252961.2 XP_006253023.1 3 - 4 391 93

View Full Product Details