Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Haus8
Entrez Gene ID
290626
Gene Name
HAUS augmin-like complex, subunit 8
Gene Aliases
Ny-sar-48, RGD1311565
Chromosome Location
Chr.16: 19654785 - 19669213 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024971.2 NP_001020142.2 8 - 9 825 93
XM_008771074.1 XP_008769296.1 9 - 10 967 93
XM_008771075.1 XP_008769297.1 9 - 10 964 93
XM_008771076.1 XP_008769298.1 9 - 10 964 93
XM_008771077.1 XP_008769299.1 8 - 9 870 93
XM_008771078.1 XP_008769300.1 5 - 6 488 93
GenBank mRNA BQ207735.1 - 3 - 4 93
FQ225627.1 - 8 - 9 839 94

View Full Product Details