Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pgls
Entrez Gene ID
290636
Gene Name
6-phosphogluconolactonase
Gene Aliases
-
Chromosome Location
Chr.16: 20020444 - 20025930 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106066.1 NP_001099536.1 4 - 5 2155 110

View Full Product Details