The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044230.2 | NP_001037695.1 | 6 - 7 | 1039 | 79 |
GenBank mRNA | DQ479969.1 | - | 6 - 7 | 1017 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044240.2 | NP_001037705.1 | 6 - 7 | 1039 | 79 |
GenBank mRNA | DQ481538.1 | - | 2 - 3 | 157 | 79 |