The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107304.1 | NP_001100774.1 | 10 - 11 | 1652 | 112 |
XM_008771106.1 | XP_008769328.1 | 10 - 11 | 1539 | 112 |