Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan14
Entrez Gene ID
306324
Gene Name
tetraspanin 14
Gene Aliases
RGD1305714
Chromosome Location
Chr.16: 17538855 - 17595960 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169127.1 NP_001162598.1 8 - 9 784 77

View Full Product Details