Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cope
Entrez Gene ID
290659
Gene Name
coatomer protein complex, subunit epsilon
Gene Aliases
-
Chromosome Location
Chr.16: 20863141 - 20873521 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106076.1 NP_001099546.1 2 - 3 393 80
XM_006252876.2 XP_006252938.1 2 - 3 407 80
GenBank mRNA BC161928.1 - 2 - 3 407 80
FQ220757.1 - 2 - 3 217 80
FQ232202.1 - 2 - 3 216 80
FQ233163.1 - 2 - 3 216 80

View Full Product Details