Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cope
Entrez Gene ID
290659
Gene Name
coatomer protein complex, subunit epsilon
Gene Aliases
-
Chromosome Location
Chr.16: 20863141 - 20873521 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252876.2 XP_006252938.1 5 - 6 715 77
GenBank mRNA FQ220757.1 - 5 - 6 525 77
FQ232202.1 - 5 - 6 524 77
FQ233163.1 - 5 - 6 524 77

View Full Product Details