Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cope
Entrez Gene ID
290659
Gene Name
coatomer protein complex, subunit epsilon
Gene Aliases
-
Chromosome Location
Chr.16: 20863141 - 20873521 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252876.2 XP_006252938.1 7 - 8 955 80
GenBank mRNA FQ220757.1 - 7 - 8 765 80
FQ232202.1 - 7 - 8 764 80

View Full Product Details