Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sugp1
Entrez Gene ID
290666
Gene Name
SURP and G patch domain containing 1
Gene Aliases
Sf4
Chromosome Location
Chr.16: 21100923 - 21131795 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011920.1 NP_001011920.1 2 - 3 223 85
GenBank mRNA BC079341.1 - 2 - 3 223 85

View Full Product Details