Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Yjefn3
Entrez Gene ID
498608
Gene Name
YjeF N-terminal domain containing 3
Gene Aliases
RGD1563236
Chromosome Location
Chr.16: 21282343 - 21288415 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_573886.6 XP_573886.4 3 - 4 410 141

View Full Product Details