The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106065.1 | NP_001099535.1 | 3 - 4 | 913 | 68 |
XM_006252856.2 | XP_006252918.1 | 2 - 3 | 1543 | 68 | |
XM_008771080.1 | XP_008769302.1 | 2 - 3 | 1262 | 68 | |
GenBank mRNA | BC169113.1 | - | 2 - 3 | 1005 | 68 |