Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lpar2
Entrez Gene ID
498609
Gene Name
lysophosphatidic acid receptor 2
Gene Aliases
Edg4, Pbx4, RGD1561336
Chromosome Location
Chr.16: 21343313 - 21348913 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109109.1 NP_001102579.1 1 - 2 104 70

View Full Product Details