Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hapln4
Entrez Gene ID
361129
Gene Name
hyaluronan and proteoglycan link protein 4
Gene Aliases
-
Chromosome Location
Chr.16: 21081377 - 21089508 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108398.1 NP_001101868.1 3 - 4 574 58

View Full Product Details