Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap1m1
Entrez Gene ID
306332
Gene Name
adaptor-related protein complex 1, mu 1 subunit
Gene Aliases
-
Chromosome Location
Chr.16: 19288354 - 19303818 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044239.1 NP_001037704.1 7 - 8 889 59
GenBank mRNA BC107903.1 - 7 - 8 889 59
FQ212761.1 - 1 - 2 50 59

View Full Product Details