The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047104.1 | NP_001040569.1 | 2 - 3 | 288 | 132 |
XM_006252908.2 | XP_006252970.1 | 2 - 3 | 936 | 132 | |
GenBank mRNA | BC098704.1 | - | 2 - 3 | 288 | 132 |