Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syap1
Entrez Gene ID
302678
Gene Name
synapse associated protein 1
Gene Aliases
-
Chromosome Location
Chr.X: 33528072 - 33562589 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004253.1 NP_001004253.1 1 - 2 360 73
XM_006256889.2 XP_006256951.1 1 - 2 373 73
GenBank mRNA BC079116.1 - 1 - 2 360 73

View Full Product Details