Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb11
Entrez Gene ID
302666
Gene Name
ankyrin repeat and SOCS box-containing 11
Gene Aliases
RGD1566298
Chromosome Location
Chr.X: 31758294 - 31781862 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106962.1 NP_001100432.1 3 - 4 308 60
XM_006256858.2 XP_006256920.1 3 - 4 662 60
XM_008773166.1 XP_008771388.1 3 - 4 713 60
GenBank mRNA FQ216453.1 - 3 - 4 414 60

View Full Product Details