Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chchd6
Entrez Gene ID
297436
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Gene Aliases
-
Chromosome Location
Chr.4: 121340952 - 121565219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106608.1 NP_001100078.1 1 - 2 183 133
XM_006236846.2 XP_006236908.1 1 - 2 190 133
GenBank mRNA FQ219980.1 - 1 - 2 164 133

View Full Product Details