The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012671.2 | NP_036803.1 | 5 - 6 | 617 | 159 |
XM_008763050.1 | XP_008761272.1 | 5 - 6 | 1511 | 159 | |
GenBank mRNA | M31076.1 | - | 5 - 6 | 617 | 159 |