Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Immt
Entrez Gene ID
312444
Gene Name
inner membrane protein, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.4: 99753452 - 99789921 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC083835.1 - - 2031 79

View Full Product Details