Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mpi
Entrez Gene ID
300741
Gene Name
mannose phosphate isomerase (mapped)
Gene Aliases
-
Chromosome Location
Chr.8: 62324176 - 62332080 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004081.1 NP_001004081.1 7 - 8 1061 86
GenBank mRNA BC079111.1 - 7 - 8 1061 86

View Full Product Details