The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007662.1 | NP_001007663.1 | 4 - 5 | 776 | 149 |
GenBank mRNA | BC081979.1 | - | 4 - 5 | 776 | 149 |