Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arcn1
Entrez Gene ID
300674
Gene Name
archain 1
Gene Aliases
-
Chromosome Location
Chr.8: 49051257 - 49075861 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007662.1 NP_001007663.1 5 - 6 946 111
GenBank mRNA BC081979.1 - 5 - 6 946 111

View Full Product Details