The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017286.2 | NP_058982.1 | 8 - 9 | 1424 | 91 |
GenBank mRNA | BC089100.1 | - | 8 - 9 | 1449 | 91 |
J05156.1 | - | 8 - 9 | 1444 | 91 | |
M22615.1 | - | 3 - 4 | 395 | 91 |