Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx22
Entrez Gene ID
300796
Gene Name
sorting nexin 22
Gene Aliases
-
Chromosome Location
Chr.8: 71725405 - 71728730 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106832.1 NP_001100302.1 5 - 6 450 148

View Full Product Details