Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smad3
Entrez Gene ID
25631
Gene Name
SMAD family member 3
Gene Aliases
Madh3, Smad 3, mad3
Chromosome Location
Chr.8: 68569530 - 68678349 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013095.3 NP_037227.1 1 - 2 409 149
GenBank mRNA BC064437.1 - - 409 149
U66479.1 - 1 - 2 218 149

View Full Product Details