Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fdxacb1
Entrez Gene ID
315646
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
RGD1562252
Chromosome Location
Chr.8: 55196474 - 55201613 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243031.2 XP_006243093.1 2 - 3 694 66

View Full Product Details