Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cilp
Entrez Gene ID
315761
Gene Name
cartilage intermediate layer protein, nucleotide pyrophosphohydrolase
Gene Aliases
-
Chromosome Location
Chr.8: 70760163 - 70775891 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108161.1 NP_001101631.1 3 - 4 305 94
XM_006243259.2 XP_006243321.1 4 - 5 460 94

View Full Product Details