Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scamp5
Entrez Gene ID
65171
Gene Name
secretory carrier membrane protein 5
Gene Aliases
-
Chromosome Location
Chr.8: 62222648 - 62248011 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031726.1 NP_113914.1 5 - 6 557 65
GenBank mRNA AF240784.1 - 5 - 6 557 65

View Full Product Details