Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Edc3
Entrez Gene ID
315708
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
Lsm16, RGD1306898
Chromosome Location
Chr.8: 62482126 - 62527665 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729908.4 XP_002729954.2 1 - 2 95 82

View Full Product Details