The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014209.1 | NP_001014231.1 | 2 - 3 | 404 | 73 |
GenBank mRNA | BC088858.1 | - | 2 - 3 | 404 | 73 |
FQ228964.1 | - | 2 - 3 | 427 | 73 |