Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arid3b
Entrez Gene ID
367092
Gene Name
AT rich interactive domain 3B (Bright like)
Gene Aliases
-
Chromosome Location
Chr.8: 62569601 - 62616837 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109001.1 NP_001102471.1 2 - 3 671 79
XM_006243171.2 XP_006243233.1 3 - 4 768 79
XM_008766323.1 XP_008764545.1 3 - 4 2915 79

View Full Product Details