Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid3b
Entrez Gene ID
367092
Gene Name
AT rich interactive domain 3B (Bright like)
Gene Aliases
-
Chromosome Location
Chr.8: 62569601 - 62616837 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109001.1 NP_001102471.1 7 - 8 1588 81
XM_006243171.2 XP_006243233.1 8 - 9 1685 81
XM_008766323.1 XP_008764545.1 8 - 9 3832 81

View Full Product Details