Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Susd1
Entrez Gene ID
298031
Gene Name
sushi domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 76514780 - 76636481 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238210.2 XP_006238272.1 13 - 14 3527 98
XM_006238211.2 XP_006238273.1 13 - 14 3524 98
XM_006238212.2 XP_006238274.1 13 - 14 1915 98
XM_006238213.2 XP_006238275.1 12 - 13 3347 98

View Full Product Details