Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsdl2
Entrez Gene ID
313200
Gene Name
hydroxysteroid dehydrogenase like 2
Gene Aliases
RGD1305387
Chromosome Location
Chr.5: 76812888 - 76848853 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025697.1 NP_001020868.1 3 - 4 395 68
XM_008763735.1 XP_008761957.1 3 - 4 438 68
XM_008763736.1 XP_008761958.1 3 - 4 437 68
GenBank mRNA BC097407.1 - 3 - 4 395 68

View Full Product Details