Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txndc8
Entrez Gene ID
362525
Gene Name
thioredoxin domain containing 8
Gene Aliases
Sptrx3
Chromosome Location
Chr.5: 75088071 - 75116427 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004092.2 NP_001004092.1 3 - 4 208 69
GenBank mRNA AY496270.1 - 3 - 4 211 69

View Full Product Details