Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgm2
Entrez Gene ID
56083
Gene Name
transglutaminase 2
Gene Aliases
TgaseII
Chromosome Location
Chr.3: 154597165 - 154627257 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019386.2 NP_062259.2 11 - 12 1831 140
GenBank mRNA AF106325.1 - 10 - 11 1782 140
BC062062.1 - 11 - 12 1831 140

View Full Product Details