Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sys1
Entrez Gene ID
685079
Gene Name
Sys1 golgi trafficking protein
Gene Aliases
-
Chromosome Location
Chr.3: 160908770 - 160921327 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001062200.5 XP_001062200.2 1 - 2 245 84

View Full Product Details