The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106541.1 | NP_001100011.1 | 3 - 4 | 602 | 65 |
XM_006235533.2 | XP_006235595.1 | 3 - 4 | 407 | 65 |