The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170666.1 | NP_733766.1 | 5 - 6 | 591 | 59 |
XM_008762471.1 | XP_008760693.1 | 5 - 6 | 708 | 59 | |
GenBank mRNA | AF548739.1 | - | 5 - 6 | 591 | 59 |