Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpm1
Entrez Gene ID
296394
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.3: 164966637 - 164986180 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106544.1 NP_001100014.1 1 - 2 189 75
GenBank mRNA FQ213909.1 - 1 - 2 167 75
FQ214139.1 - 1 - 2 172 75
FQ216824.1 - 1 - 2 172 75
FQ219919.1 - 1 - 2 172 75

View Full Product Details