Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wfdc2
Entrez Gene ID
286888
Gene Name
WAP four-disulfide core domain 2
Gene Aliases
re4
Chromosome Location
Chr.3: 161018497 - 161036000 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173109.1 NP_775132.1 2 - 3 264 58
XM_008762473.1 XP_008760695.1 2 - 3 269 58
GenBank mRNA AJ515239.1 - 2 - 3 264 58

View Full Product Details