Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ghrh
Entrez Gene ID
29446
Gene Name
growth hormone releasing hormone
Gene Aliases
GHRF
Chromosome Location
Chr.3: 153449124 - 153468794 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031577.1 NP_113765.1 1 - 2 179 58
GenBank mRNA M73486.1 - 1 - 2 179 58

View Full Product Details