Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ghrh
Entrez Gene ID
29446
Gene Name
growth hormone releasing hormone
Gene Aliases
GHRF
Chromosome Location
Chr.3: 153449124 - 153468794 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762304.1 XP_008760526.1 1 - 2 81 80
GenBank mRNA U10156.1 - 1 - 2 81 80
Z34092.1 - 1 - 2 81 80

View Full Product Details