Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dbndd2
Entrez Gene ID
499941
Gene Name
dysbindin (dystrobrevin binding protein 1) domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 160934082 - 160938468 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235610.1 XP_006235672.1 1 - 2 312 102
GenBank mRNA FQ220039.1 - 1 - 2 137 102

View Full Product Details