The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199392.2 | NP_955424.2 | 5 - 6 | 910 | 60 |
XM_006235578.2 | XP_006235640.1 | 5 - 6 | 944 | 60 | |
GenBank mRNA | AB096685.1 | - | 5 - 6 | 911 | 60 |