The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012093.1 | NP_001012093.1 | 3 - 4 | 531 | 70 |
GenBank mRNA | BC085820.1 | - | 3 - 4 | 531 | 70 |