Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Matn4
Entrez Gene ID
296358
Gene Name
matrilin 4
Gene Aliases
-
Chromosome Location
Chr.3: 160838632 - 160853650 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106539.1 NP_001100009.1 8 - 9 1720 104
XM_006235531.2 XP_006235593.1 8 - 9 1629 104

View Full Product Details